Natriumhydrogencarbonat 75 g/l in wässriger Lösung
Catalog # EURLECM0980D
Supplier: EUROCLONE
CAS Number:
Specifications
- Packung:Glass bottle
- VE:100 mL
- Seq:2440
Specifications
About this item
:
:84.01 g/mol
Storage Temperature:Raumtemperatur
EINECS:205-633-8
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528