Bovine serum albumin (BSA), Fraction V
- CAS Nr.:90604-29-8
9048-46-8 - Ursprung:Serum
- Spezies:Bovine
- Lagerbedingungen:Refrigerator
- Proteinsynonym:BSA|Fraction V|Cohn Fraction V
- Protein/Peptid-Name:Bovine Serum Albumin
- pH:pH 5.2
Frequently Bought Together