Digital rotational viscometers, DV-I Prime
: AMETEK
The only viscometer in its class to offer continuous sensing and data display.
- RS-232 PC interface for use with optional wingather software
- Analog outputs for recording torque and temperature
- 18 speeds provide great range capability
- Direct access to time measurement function
- Output connection to printer
- Scroll Keys for simple selection of speed and spindle
- Displayed information includes % torque, speed/spindle and temperature (°C or °F) if optional RTD temperature probe is installed.
Kegel-/Plattenmodelle sind ebenfalls erhältlich.
Temperature off-set capability to ±1 °C.
: Wingather software, RTD temperature probe , ball bearing suspension (RV/HA/HB), viscosity standards, RV/HA/HB-1 spindle, quick action lab stand, EZ-lock spindle coupling system, temperature bath, small sample adapter, UL adapter, thermosel, helipath stand with T-bar spindles, spiral adapter, DIN adapter, quick connect/extension links, vane spindles and protective keypad covers can be ordered separately.
: Includes instrument, six spindles (RV/HA/HB) or four spindles (LV), spindle guard leg, lab stand (Model S) and carrying case.
Frequently Bought Together