Oxygen meter, online, insert, Quadroline Oxi 296
Catalog # 664-0015
Supplier: WTW ONLINE
Specifications
- Bezeichnung:Online-Sauerstoffmessgerät, in Gehäuse, Oxi 296
- VE:1
Specifications
About this item
Suitable for the most common applications in the water and waste water sectors.
- With two relay contacts and an analogue output for oxygen and temperature readings
Ordering information: Electrodes with various cable lengths, frames and mounting kits for horizontal and vertical ducts available on request.