Cover glass forceps
160 mm
Katalognr. 100-0331
Anbieter: HAMMACHER KARL
Direkt zu
Product Details & Documents
WIRONIT steel (18/12 stainless steel with 18% chrome and from 10 - 12% nickel)
- Resistant to corrosion
- Anti-magnetic
Technische Daten
- Länge:160 mm