Messkolben, Klasse A
Supplier: BDH Prolabo
Certificates
About this item
DURAN®, Borosilikatglas, mit PP-Stopfen.
- Calibrated for 'ln' capacity
- Blue embossed marking and inscriptions
- Certificate of conformity with dated batch number
DIN 12 644 T1
Certifications: DIN 12 644 T1
Frequently Bought Together