Desiccator Cabinets
Polystyrol-Exsikkator, Türöffnung 188×110 (mm), 200×225×168 mm
Katalognr. 467-0004
Anbieter: SciLabware
Direkt zu
Product Details & Documents
Clear, high impact polystyrene cabinets have an airtight door with foam rubber gasket.
- Cabinets can be stacked to save space
: Supplied with sealed jar of silica gel.
Technische Daten
- Außen B×T×H:200×225×168 mm
- Bezeichnung:Polystyrol-Exsikkator
- Mit:Türöffnung 188×110 (mm)