Tubing clips
Katalognr. 229-0041
Anbieter: ALPACO
Technische Daten
- Spannweite:14 - 19 mm
- VE:10
Technische Daten
Über diesen Artikel
Nickel-chromium steel strip. With brass screw and galvanized steel hexagonal head. Safety tube clips for attaching tubes to pipes and fixtures.
- Corrosion resistant, very good chemical resistance
- Quick, easy assembly and disassembly
- Extremely durable