TWEEN® 20 Detergent, Molecular Biology Grade
Catalog # MILF655204-100MLCN
Supplier: Merck
Specifications
- Bezeichnung:TWEEN® 20 detergent, molecular biology grade
- Environmentally Preferable:
- Lagertemperatur:15...30 °C
- VE:100 mL
Specifications
About this item
Detergent, molecular biology grade - CAS 9005-64-5 - Calbiochem