Flask and Neoprene Stopper
N° de catalogue MILFXX6602501
Fournisseur: Merck
Spécifications
- Bezeichnung:Flask and neoprene stopper, borosilicate glass, neoprene bulb and stopper
- VE:1
Spécifications
À propos de cet article
Simultaneously vacuum filter 12 samples for rapid analysis of retentate or filtrate. Numbered positions on the filter plate assure accurate sample tracking. Sealing stoppers maintain vacuum on unused sample cups. Accessory extension barrels increase sample volumes from 15 to 50 ml.
General filtration of 15 to 50 ml samples, preparation for scintillation counting.