Plastic Crate with Lid
Grau, 31 L, PP, 400×300×333 mm
Katalognr. LAMA693
Anbieter: LAMAPLAST
Direkt zu
Product Details & Documents
Crates have one handle on the longest side, this is placed centrally. Crates are light yet strong, providing an economical alternative to metal crates.
- Polypropylene
- Colour: Grey
- For food use
Dokumente
TDS Plastic Crate with Lid, 31 L
Technisches Datenblatt
Technische Daten
- Kapazität:31 L
- Farbe:Grau
- Material:PP
- B×T×H:400×300×333 mm