ContactUsLinkComponent
Accessories for EasySampler Probe 210 Set
Katalognr. METB30247094
Anbieter:  Mettler - Toledo
New Product
Accessories for EasySampler Probe 210 Set
Katalognr. METB30247094
Anbieter:  Mettler - Toledo
New Product
Technische Daten
  • Bezeichnung:
    Tube set probe 210, PTFE tube, PEEK fittings; 4 pcs in a set
  • VE:
    1