Organischer Referenzstandard, Phenol 100 µg/ml in Acetonitril, Supelco®
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338
Specifications
- CAS Nr.:108-95-2
- Lagerbedingungen:2...8 °C
- Beschreibung Gefahrenklasse:H225 - H302 + H312 + H332 - H319
- Shipping conditions:STD
- MDL No.:MFCD00002143
- M-Clarity Qualitätsstufe:MQ100
- Formel:C₆H₅OH
- Molecular weight:94,11
- Flammpunkt:2 °C - closed cup