X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Catalog # MOLE11322533-5G
Supplier: Molekula
CAS Number:
Specifications
- Packung:Glass bottle for solids
- VE:5 g
- Seq:3173
Specifications
About this item
:
:408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Gefrierschrank
EINECS:230-640-8
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666