EP Reference Solution BY2, Brownish-Yellow Reference Standard
EP Reference Solution BY2, Brownish-Yellow Standard Solution, Glasflasche, braun, 120 mL
Katalognr. RICC28724
Anbieter: RICCA CHEMICAL
EP Reference Solution BY2, Brownish-Yellow Reference Standard
Katalognr. RICC28724
Anbieter: RICCA CHEMICAL
CAS Nummer:
Direkt zu
Product Details & Documents
Technische Daten
Technische Daten
- Packung:Glasflasche, braun
- VE:120 mL
- Bezeichnung:EP Reference Solution BY2, Brownish-Yellow Standard Solution
- Cat. No.:RICC28724
- Aussehen:Brownish-yellow liquid