Order Entry
Switzerland
ContactUsLinkComponent
BIOPHEN™ CS-31(02) Kallikrein Chromogenic Substrate
Katalognr. CMPP229031
New Product
CAS-Nummer:  
BIOPHEN™ CS-31(02) Kallikrein Chromogenic Substrate
Katalognr. CMPP229031
New Product
CAS-Nummer:  

Technische Daten

  • VE:
    25 mg
  • Lagerbedingungen:
    2...8 °C
  • Molecular weight:
    538,6 Da
  • Sequence:
    D-Pro-Phe-Arg-pNa, 2HCl
  • Molecular formula:
    C₂₆H₃₄N₈O₅,₂HCl

Technische Daten

Über diesen Artikel

Synthetic generic chromogenic substrate, highly purified and stabilised. Recommended substrate for Kallicrein.

  • Molarity: about 46 μmol/vial
  • Free pNA content (measured on substrate at 2,5 g/L): Ext.Ø 405 nm ≤0,30
  • Purity grade: >95%
  • pNA content (measured on RP-HPLC): RP-HPLC <0,1%

Presentation: Vial containing 25 mg of BIOPHEN™ CS-31(02), lyophilised in presence of Mannitol as a bulking agent.

According to the research protocol used, the BIOPHEN™ CS-31(02) chromogenic substrate can be restored with variable volumes of distilled water; for example 5 ml can be used for a substrate concentration of 5 mg/ml, or 20 ml for a substrate concentration of 1,25 mg/ml.

Caution: This kit is for research use only and should not be used for patient diagnosis or treatment. Avoid exposure to light.