AuftragsverfolgungKontakt
Gas Chromatography Mass Spectrometry (GCMS) Systems | Avantor
Gas Chromatography Mass Spectrometry (GCMS) Systems
Katalognr.
Gas Chromatography Mass Spectrometry (GCMS) Systems
Katalognr.
Anbieter:  
Gas Chromatography Mass Spectrometry (GCMS) Systems
Gas Chromatography Mass Spectrometry (GCMS) Systems
Product Details & Documents
Agilent GC/MS systems combine the power of gas chromatography with the precision of mass spectrometry to deliver high-sensitivity, high-throughput analysis across a wide range of applications. With over 40 years of innovation, Agilent GC/MS platforms are trusted worldwide for their performance, reliability, and ease of use.
  • Comprehensive portfolio: Single quad, triple quad, and Q-TOF systems
  • High Efficiency Source (HES) for ultratrace-level detection
  • HydroInert source for hydrogen carrier gas compatibility
  • MassHunter software for intuitive data acquisition and analysis
  • Application-specific analyzers for environmental, food, pharma, and forensics
  • Robust ion source technology for extended uptime and reduced maintenance
  • Preconfigured libraries and methods for fast startup and method development
  • Flexible detection modes: Full scan, SIM, MS/MS, and HRAM
Models: 5977C GC/MSD, 7000E/7010D GC/TQ, 7250 GC/Q-TOF.
Mass range: Up to 1,700 m/z (model dependent).
Ionization modes: EI, CI, low-energy EI, MS/MS.
Mass resolution: High-resolution accurate mass (HRAM) with Q-TOF.
Carrier gas compatibility: Helium and hydrogen.
Software: MassHunter suite for acquisition, quantitation, and reporting.
Applications: Routine testing, research, trace-level detection, and regulatory compliance.
The 7010C GC triple quadrupole mass spectrometry system is the most sensitive instrument of the Agilent GC/TQ portfolio, built on an intelligent platform to maximize productivity by reducing unplanned instrument downtime. With downtime minimized, the 7010C GC/TQ can lower your lab’s environmental impact while reaching ultralow limits of detection down to the attogram level.
The 7250 GC/Q-TOF system delivers full-spectrum, high-resolution, accurate mass data with a wide dynamic range for identifying and quantifying GC-amenable compounds. This high-resolution GC/Q-TOF enables accurate mass screening by GC/MS and enhanced compound identification through MS/MS, low-energy electron ionization, and complimentary chemical ionization techniques.
Bestellinformationen: Each model is configurable for your needs. Contact your Avantor representative for more information.