JUN Dye qPCR Calibration Solution *10000X*
Katalognr. AATB67038
Anbieter: AAT Bioquest
New Product
CAS-Nummer:
Technische Daten
- VE:100 µl
- Bezeichnung:JUN Dye qPCR calibration solution *10000X*
Technische Daten
Über diesen Artikel
JUN dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.