HIS Lite™ iFluor® 647 Tris NTA-Ni Complex
Codice catalogo AATB12618
Nome del fornitore: AAT BIOQUEST
New Product
Numero CAS:
Specifiche
- VE:100 µG
- Bezeichnung:HIS Lite™ iFluor® 647 Tris NTA-Ni complex
Specifiche
Informazioni su questo articolo
iFluor 647-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.