ContactUsLinkComponent
Amplite® Colorimetric Lipase Activity Assay Kit
Catalog # AATB11315
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Amplite® Colorimetric Lipase Activity Assay Kit
Catalog # AATB11315
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • VE:
    100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® colorimetric lipase activity assay kit offers a straightforward and efficient protocol for quantifying lipase activity.
  • Robust protocol
  • Suitable for detecting lipase activity in a variety of biological samples, including cells, supernatant, tissue extracts, and serum
This kit utilises a coupled enzymatic reaction, which converts the substrate into a colored product (586 nm) proportional to the lipase activity present.
Delivery: Shipped with blue ice.
Caution: Research use only.