Amplite® Colorimetric Gama-Glutamyltransferase (GGT) Activity Assay Kit
Catalog # AATB11801
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- VE:100 Tests
- Tests pro Kit:100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Colorimetric Gamma-Glutamyltransferase (GGT) Activity Assay Kit provides an efficient and straightforward method for measuring GGT activity across various sample types.
- Robust protocol
- Absorbance measured at 418 nm
- Six-month shelf life
It is compatible with a wide range of biological samples, including cells, tissue extracts, and serum, and is suitable for high-throughput systems.
Delivery: Shipped with blue ice.
Caution: Research use only.