AuftragsverfolgungKontakt
FITC AML-NHS ester, ReadiCleave™
FITC AML-NHS ester, ReadiCleave™
1 mg
Katalognr. AATB7004
Anbieter:   AAT Bioquest
FITC AML-NHS ester, ReadiCleave™
Katalognr. AATB7004
Anbieter:  AAT Bioquest
CAS Nummer:  
FITC AML-NHS ester, ReadiCleave™
1 mg
CAS Nummer:  
Preis
583.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 mg
Product Details & Documents
ReadiCleave™ AML FITC contains an azidomethyl linker that can be cleaved with TCEP to remove the fluorescein fluorophore from the target molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Caution: For Research Use Only
  • Molekulargewicht:966.98 g/mol
Technische Daten
Technische Daten
  • VE:
    1 mg
Technische Daten