Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

mFluor™ Violet 545 SE

1 mg
Katalognr. AATB1157
Anbieter:   AAT Bioquest
mFluor™ Violet 545 SE
Katalognr. AATB1157
Anbieter:  AAT Bioquest
CAS Nummer:  
mFluor™ Violet 545 SE

1 mg

CAS Nummer:  
Preis
SFr. 290.00
Einloggen für Verfügbarkeit
1 * 1 mg
Product Details & Documents

mFluor™ Violet 545 dyes have fluorescence excitation and emission maxima of ~405 nm and ~530 nm respectively.

  • Stable for 12 months

mFluor™ dyes are developed for multicolor flow cytometry-focused applications.

 : For Research Use Only

  • Molekulargewicht:983.09 g/mol
Technische Daten

Technische Daten

  • VE:
    1 mg
  • Cat. No.:
    AATB1157

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.