Order Entry
Switzerland
ContactUsLinkComponent
Anti-SCAMP2 / SC2 Rabbit Polyclonal Antibody
Anti-SCAMP2 / SC2 Rabbit Polyclonal Antibody
Catalog # ANTIA9447-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-SCAMP2 / SC2 Rabbit Polyclonal Antibody
Catalog # ANTIA9447-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    SCAMP2 / SC2
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# O15127
  • Lagerungspuffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    38 kDa
  • Sequence:
    MSAFDTNPFADPVDVNPFQDPSVTQLTNAPQGGLAEFNPFSETNAATTVPVTQLPGSSQPAVLQPSVEPTQPTPQAVVSAAQAGLLRQQEELDRKAAELERKERELQNTVANLHVRQNNWPPLPSWCPVKPCFYQDFSTEIPADYQRICKML
  • Lagertemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Transporttemperatur:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-152 of human SCAMP2 (NP_005688.2)
  • Reinigung:
    Affinity purification
  • Packung:
    Plastic vial
  • VE:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to SCAMP2 / SC2 for WB with samples derived from Human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:100

Type: Primary
Antigen: SCAMP2 / SC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human