Anti-PKC delta + PKC theta Rabbit Polyclonal Antibody
Katalognr. ANTIA94472-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:ALPS3
- Antigen-Symbol:PKC delta + PKC theta
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# Q05655
- Antigen-Synonyme:PRKCT|CVID9|nPKC-theta|MAY1|nPKC-delta|PRKCQ|PKCD|PKC delta + PKC theta|PRKCD
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:77 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PKC delta around the phosphorylation site of Ser645 (amino acids 612-661)
- Validierte Anwendungen:IHC
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Packung:Plastic vial
- VE:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody to PKC delta (phospho Ser645) for WB, IHC and ELISA with samples derived from Human.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: PKC delta + PKC theta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human