Anti-pan-AKT Rabbit Polyclonal Antibody
Catalog # ANTIA94467-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:AKT1
- Antigen-Symbol:pan-AKT
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P31749
- Antigen-Synonyme:AKT2|AKT1_HUMAN|PRKBA|PKB|PKB beta|AKT3|PKB alpha|pan-AKT|PKBalpha
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:55 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Akt1 around the phosphorylation site of Thr450 (amino acids 416-465)
- Validierte Anwendungen:IHC
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Packung:Plastic vial
- VE:100 µG
Specifications
About this item
Rabbit polyclonal antibody to Akt1 (phospho Thr450) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:1000
Type: Primary
Antigen: pan-AKT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat