Anti-Bcl2 alpha Rabbit Polyclonal Antibody
ANTIA94462-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Apoptosis regulator Bcl2
- Antigen-Symbol:Bcl2 alpha
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P10415
- Antigen-Synonyme:Bcl 2|B-cell leukemia/lymphoma 2|B-cell CLL/lymphoma 2|B cell lymphoma protein 2 alpha isoform|Leukemia /lymphoma, B-cell, 2|Oncogene B-cell leukemia 2|Bcl2 alpha|B cell lymphoma protein 2
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:26 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BCL-2 around the phosphorylation site of Ser87 (amino acids 61-110)
- Validierte Anwendungen:IHC
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Packung:Plastic vial
- VE:100 µG
Rabbit polyclonal antibody to BCL-2 (phospho Ser87) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:1000
Type: Primary
Antigen: Bcl2 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat