Anti-p27 KIP 1 Rabbit Polyclonal Antibody
ANTIA94597-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:AA408329
- Antigen-Symbol:p27 KIP 1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P46527
- Antigen-Synonyme:CDKN 1B|CDKN1B|CDKN4|Cyclin dependent kinase inhibitor p27|AI843786|Cyclin Dependent Kinase Inhibitor 1B|CDKN 4|CDN1B_HUMAN|Cdki1b
- Lagerungspuffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:22 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human p27 Kip1 around the phosphorylation site of Thr187 (amino acids 149-198)
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Packung:Plastic vial
- VE:100 µG
Rabbit polyclonal antibody to p27 Kip1 (phospho Thr187) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: p27 KIP 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat