Anti-Calmodulin 3 Rabbit Polyclonal Antibody
ANTIA94547-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:CALM
- Antigen-Symbol:Calmodulin 3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P62158
- Antigen-Synonyme:CAM2|CAMB|CALML2|CALM2|CAM3|CAM|CAM1|Calmodulin 3|Calmodulin 3 (phosphorylase kinase, delta)
- Lagerungspuffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:16 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human Calmodulin (amino acids 46-95)
- Validierte Anwendungen:IHC
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Packung:Plastic vial
- VE:100 µG
Rabbit polyclonal antibody to Calmodulin for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: Calmodulin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat