Order Entry
Switzerland
ContactUsLinkComponent
Anti-Integrin alpha 2+beta 1 Rabbit Polyclonal Antibody
Catalog # ANTIA97457-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Integrin alpha 2+beta 1 Rabbit Polyclonal Antibody
Catalog # ANTIA97457-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CD29
  • Antigen-Symbol:
    Integrin alpha 2+beta 1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# P17301
  • Antigen-Synonyme:
    GPIa|Collagen receptor|Integrin beta 1|Integrin alpha 2+beta 1|CD49B|GPIIA|Integrin alpha 2|ITGB1|ITGA2
  • Lagerungspuffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    130 kDa
  • Lagertemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transporttemperatur:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from the C-terminal region of human ITGA2 (amino acids 1081-1130)
  • Reinigung:
    The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
  • Packung:
    Plastic vial
  • VE:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to ITGA2 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Integrin alpha 2+beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat