Anti-p53 Rabbit Polyclonal Antibody
N° de catalogue ANTIA96483-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Antikörper-Typ:Primär
- Antigen-Name:Antigen NY-CO-13
- Antigen-Symbol:p53
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P04637
- Antigen-Synonyme:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Lagerungspuffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:43 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transporttemperatur:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human p53 around the phosphorylation site of Ser46 (amino acids 11-60)
- Reinigung:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Packung:Plastic vial
- VE:100 µG
Spécifications
À propos de cet article
Rabbit polyclonal antibody to p53 (phospho Ser46) for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human