Anti-Centaurin beta 2 Goat Polyclonal Antibody
ANTIA83973-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:ACAP 2
- Antigen-Symbol:Centaurin beta 2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Goat
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen-Synonyme:ACAP2_HUMAN|ANK repeat and PH domain-containing protein 2|Arf-GAP with coiled-coil|Centaurin beta 2|CENT B 2|Arf GAP with coiled coil ANK repeat and PH domain containing protein 2|ArfGAP with coiled coil ankyrin repeat and PH domains 2|ACAP2|Arf GAP with coiled coil ANK repeat and PH domains 2
- Lagerungspuffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-EKLNRFQQDSQKF
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:100 µg at 0,5 mg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ACAP2 (C terminal)
- Validierte Anwendungen:IHC
- Reinigung:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Packung:Plastic vial
- VE:100 µG
Goat polyclonal antibody to Centaurin beta 2 for ELISA, WB and IHC with samples derived from human, mouse and rat.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: Centaurin beta 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat