Anti-ZNF202 Mouse Monoclonal Antibody [clone: PCRP-ZNF202-1C4]
Katalognr. ANTIA315799-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Zinc finger protein 202
- Antigen-Symbol:ZNF202
- Klonalität:Monoklonal
- Klon:PCRP-ZNF202-1C4
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG2a
- Reaktivität:Human
- Form:Liquid
- Gen-ID:UniprotID# O95125
- Antigen-Synonyme:ZNF 202|ZKSCAN 10|ZNF202|ZKSCAN10|Zinc finger protein with KRAB and SCAN domains 10|ZN202_HUMAN
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:74,72 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:200 µg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 37-132) of human ZNF202 protein
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Technische Daten
Über diesen Artikel
Mouse monoclonal [PCRP-ZNF202-1C4] antibody to ZNF202 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: ZNF202
Clonality: Monoclonal
Clone: PCRP-ZNF202-1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human