Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human STAT6 ELISA Kit | Avantor

Human STAT6 ELISA Kit

Human STAT6 ELISA kit, 96 Tests
Katalognr. ANTIA310566-96
Anbieter:   Antibodies.com
Human STAT6 ELISA Kit
Katalognr. ANTIA310566-96
Anbieter:  Antibodies.com
CAS Nummer:  
Human STAT6 ELISA Kit
Human STAT6 ELISA Kit

Human STAT6 ELISA kit, 96 Tests

CAS Nummer:  
Preis
SFr. 564.00
Einloggen für Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Human STAT6 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human STAT6 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7,98 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human STAT6 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human STAT6 in An antibody specific for STAT6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the STAT6 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-STAT6 antibody, which also binds the STAT6 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-STAT6 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-STAT6 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of STAT6 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of STAT6 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestellinformationen: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Technische Daten

Technische Daten

  • VE:
    96 Tests
  • Bezeichnung:
    Human STAT6 ELISA kit
  • Empfindlichkeit:
    7,98 ng/L
  • Reaktivität der primären Antikörper:
    Human
  • Shelf life:
    Store for 6 months at 4 °C
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA310566-96
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Format:
    Pre-coated
  • Target protein:
    STAT6
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erfassungsbereich:
    100 - 1600 ng/L
  • Regulatory status:
    RUO

Technische Daten

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..