Mouse PGK1 ELISA Kit
Product Details & Documents
Mouse PGK1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PGK1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 200 to 3200 ng/L
- Sensitivity: 9,04 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse PGK1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PGK1 in serum, plasma or other biological fluids. An antibody specific for PGK1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PGK1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-PGK1 antibody, which also binds the PGK1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-PGK1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-PGK1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of PGK1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of PGK1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- VE:96 Tests
- Bezeichnung:Mouse PGK1 ELISA kit
- Empfindlichkeit:9,04 ng/L
- Reaktivität der primären Antikörper:Mouse
- Shelf life:Store for 6 months at 4 °C
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ANTIA311267-96
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Format:Pre-coated
- Target protein:PGK1
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, plasma or other biological fluids
- Erfassungsbereich:200 - 3200 ng/L
- Regulatory status:RUO
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



