Mouse eNOS ELISA Kit
Product Details & Documents
Mouse eNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse eNOS in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection Range: 15,625 to 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse eNOS ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse eNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for eNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the eNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-eNOS Antibody, which binds the captured eNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of eNOS captured in each well. The concentration of eNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Bestellinformationen: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Technische Daten
Technische Daten
- VE:96 Tests
- Bezeichnung:Mouse eNOS ELISA kit
- Testprinzip:Quantitative
- Empfindlichkeit:9,375 pg/ml
- Reaktivität der primären Antikörper:Mouse
- Shelf life:Store for 6 months at 4 °C
- Testdauer:Multiple steps
- Time to Results:4 h 30 min
- Cat. No.:ANTIA74103-96
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Format:Pre-coated
- Target protein:eNOS
- Kreuzreaktivität:Mouse eNOS ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse eNOS. No significant cross-reactivity or interference between eNOS and analogues was observed.
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, plasma, tissue homogenates and other biological fluids
- Erfassungsbereich:15,625 - 1000 pg/ml
- Regulatory status:RUO
Technische Daten
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



