Order Entry
Switzerland
ContactUsLinkComponent
Porcine Cardiac Troponin T ELISA Kit
Katalognr. ANTIA73865-96
Anbieter:  ANTIBODIES.COM
New Product
CAS-Nummer:  
undefined
Porcine Cardiac Troponin T ELISA Kit
Katalognr. ANTIA73865-96
Anbieter:  ANTIBODIES.COM
New Product
CAS-Nummer:  

Technische Daten

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Other
  • Target protein:
    Cardiac Troponin T
  • Bezeichnung:
    Porcine cardiac troponin T ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kreuzreaktivität:
    Porcine Cardiac Troponin T ELISA Kit exhibits high specificity and excellent specificity for the detection of porcine Cardiac Troponin T. No significant cross-reactivity or interference between Cardiac Troponin T and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Testprinzip:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    15,625 - 1000 pg/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    9,375 pg/ml
  • Regulatory status:
    RUO

Technische Daten

Über diesen Artikel

Porcine Cardiac Troponin T ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Cardiac Troponin T in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 15,625 to 1000 pg/ml
  • Sensitivity: 9,375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Porcine Cardiac Troponin T ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Cardiac Troponin T in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cardiac Troponin T has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cardiac Troponin T present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cardiac Troponin T Antibody, which binds the captured Cardiac Troponin T present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cardiac Troponin T captured in each well. The concentration of Cardiac Troponin T can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.