Order Entry
Switzerland
ContactUsLinkComponent
Human PGC1 alpha ELISA Kit
Human PGC1 alpha ELISA Kit
Codice catalogo ANTIA74512-96
Nome del fornitore:  ANTIBODIES.COM
New Product
Numero CAS:  
undefined
Human PGC1 alpha ELISA Kit
Codice catalogo ANTIA74512-96
Nome del fornitore:  ANTIBODIES.COM
New Product
Numero CAS:  

Specifiche

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    PGC1 alpha
  • Bezeichnung:
    Human PGC1 alpha ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kreuzreaktivität:
    Human PGC1 alpha ELISA kit exhibits high specificity and excellent specificity for the detection of human PGC1 alpha. No significant cross-reactivity or interference between PGC1 alpha and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Testprinzip:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    0,313 - 20 ng/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    0,188 ng/ml
  • Regulatory status:
    RUO

Specifiche

Informazioni su questo articolo

Human PGC1 alpha ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PGC1 alpha in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human PGC1 alpha ELISA kit (A74512) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PGC1 alpha in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PGC1 alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PGC1 alpha present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PGC1 alpha Antibody, which binds the captured PGC1 alpha present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PGC1 alpha captured in each well. The concentration of PGC1 alpha can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.