Human CD2 ELISA Kit
Product Details & Documents
Human CD2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0,313 - 20 ng/ml
- Sensitivity: 0,188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human CD2 ELISA kit (A79499) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CD2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CD2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CD2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CD2 Antibody, which binds the captured CD2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CD2 captured in each well. The concentration of CD2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Technische Daten
Technische Daten
- VE:96 Tests
- Bezeichnung:Human CD2 ELISA kit
- Testprinzip:Quantitative
- Empfindlichkeit:0,188 ng/ml
- Reaktivität der primären Antikörper:Human
- Shelf life:Store for 6 months at 4 °C
- Testdauer:Multiple steps
- Time to Results:4 h 30 min
- Cat. No.:ANTIA79499-96
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Format:Pre-coated
- Target protein:CD2
- Kreuzreaktivität:Human CD2 ELISA kit exhibits high specificity and excellent specificity for the detection of human CD2. No significant cross-reactivity or interference between CD2 and analogues was observed.
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, plasma, tissue homogenates and other biological fluids
- Erfassungsbereich:0,313 - 20 ng/ml
- Regulatory status:RUO
Technische Daten
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



