ContactUsLinkComponent
Crimp Vial, Quartz Glass
  NEUB7820-QU-04
New Product
Crimp Vial, Quartz Glass
  NEUB7820-QU-04
New Product
  • Mit Deckel:
    Nein
  • Ø×H:
    15×45 mm
  • Kapazität:
    4,00 ml
  • Bezeichnung:
    Crimp vials made of quartz glass
  • Material:
    Quartz
  • VE:
    1
Crimp vial made of quartz glass.
  • Without cap
  • Without labeling