Anti-Syndecan-1 Mouse Monoclonal Antibody [clone: B-A38]
ABCAAB714-500
: Abcam
- Antikörper-Typ:Primär
- Antigen-Name:Syndecan-1
- Antigen-Symbol:SDC1
- Klonalität:Monoklonal
- Klon:B-A38
- Konjugation:Unconjugated
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG1
- Reaktivität:
- Environmentally Preferable:
- Form:Liquid
- Antigen-Synonyme:SDC|SDC1|SYND1
- Lagerungspuffer:pH: 7,3
Constituents: PBS, Tissue culture supernatant - Lagertemperatur:−20...+4 °C
- Transporttemperatur:20
- VE:500 µl
Mouse Monoclonal Syndecan-1 antibody. Carrier free. Suitable for IHC-P, IHC-Fr and reacts with Human samples.
Type: Primary
Antigen: Syndecan-1
Clonality: Monoclonal
Clone: B-A38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: