Thiamin (Vitamin B1)
Catalog # CHMCAB5775
Supplier: Chemicon
CAS Number:
Specifications
- VE:100 µl
- Seq:1286
Specifications
About this item
:
Merck Index:12,09430
Boiling Pt:122…128 °C (0.03 mmHg)
Melting Pt:128…129 °C
Density:1.43 g/cm³ (20 °C)
:300.81 g/mol
EINECS:200-425-3
CAS Number:59-43-8
Formula:C₁₂H₁₇ClN₄OS
MDL Number:MFCD00044586