Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 840)
Anbieter: AAT BIOQUEST
Zertifikate
Über diesen Artikel
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Technische Daten
- Catalog No:
- AATB100330Q1
- AATB100330Q0
- Antigen-Symbol:
- CD3e
- CD3e
- Antigen-Name:
- CD3e antigen
- CD3e antigen
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- iFluor® 840
- iFluor® 840
- Klon:
- SK7
- SK7
- Reaktivität:
- Human
- Human
- Wirt:
- Mouse
- Mouse
- ImmunFluoreszenz:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Kreuzadsorption:
- No
- No
- Lagertemperatur:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Transporttemperatur:
- 20
- 20
Technische Daten
Frequently Bought Together