Specifications
- Antikörper-Typ:Primär
- Antigen-Name:SRY (sex determining region Y)-box 2
- Antigen-Symbol:SOX2
- Klonalität:Monoklonal
- Klon:SOX2/1791
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG2b kappa
- Reaktivität:Human
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gen-ID:P48431
- Antigen-Synonyme:SOX2
- Lagerungspuffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Lagertemperatur:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Konzentration:0,2 mg/ml
- Transporttemperatur:4 °C
- Immunogen:Amino acids 176-305 of the human protein were used as the immunogen for the SOX-2 antibody.
- Reinigung:Protein G affinity chromatography
- Größe:100 µg
- VE:100 µG
Specifications
About this item
SOX2 is a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. At present, 30 Sox genes have been identified. SOX2 is required for stem cell maintenance in the central nervous system, and it also regulates gene expression in the stomach. SOX2 is necessary for regulating multiple transcription factors that affect Oct 3/4 expression. An essential function of SOX2 is to stabilize embryonic stem cells in a pluripotent state by maintaining the requisite level of Oct 3/4 expression.
ELISA: 2-4 µg/ml; order BSA-free formulation for coating Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT Optimal dilution of the SOX-2 antibody should be determined by the researcher.
Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: SOX2/1791
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human