Anti-Prkcb Rabbit Monoclonal Antibody [clone: G2]
Catalog # PRSI54-330
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:protein kinase C, beta
- Antigen-Symbol:Prkcb
- Klonalität:Monoklonal
- Klon:G2
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Western blot:Yes
- Gen-ID:18751
- Antigen-Synonyme:PKC-Beta|Prkcb2|PKC-B|Pkcb|Prkcb1
- Modification:None
- Lagerungspuffer:PBS, pH 7,2, 0,02% NaN₃
- Lagertemperatur:Store at 4 °C for up to 6 months. Store at −20 °C long term
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Synthetic peptide derived from mouse Prkcb (DGVTTKpTFCGTPDYI)
- Reinigung:Protein A purified
- Packung:Vial
- VE:100 µG
Specifications
About this item
Type: Primary
Antigen: Prkcb
Clonality: Monoclonal
Clone: G2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse