Anti-RAB2A Rabbit Polyclonal Antibody
Katalognr. PRSI14-077
Anbieter: ProSci Inc.
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:RAB2A, member RAS oncogene family
- Antigen-Symbol:RAB2A
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:5862
- Antigen-Synonyme:RAB2|RAB2A|member RAS oncogene family
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human RAB2A (NP_002856.1)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: RAB2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat