Anti-DDR1 Rabbit Polyclonal Antibody
Catalog # PRSI13-682
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:discoidin domain receptor tyrosine kinase 1
- Antigen-Symbol:DDR1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:780
- Antigen-Synonyme:NEP|HGK2|MCK10|CD167|Cell adhesion kinase|Protein-tyrosine kinase 3A|DDR1|Discoidin domain receptor 1|NTRK4|EDDR1|CAK|MCK-10|RTK6|ENTRK4|Tyrosine-protein kinase CAK|Mammary carcinoma kinase 10|PTK3|CD167a antigen|Protein-tyrosine kinase RTK-6|PTK3A|TRK E|Tyrosine kinase DDR|DDR|TRKE
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 290-420 of human DDR1 (NP_054700.2)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: DDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human