Anti-HSPD1 Rabbit Polyclonal Antibody
PRSI61-556
- Antikörper-Typ:Primär
- Antigen-Name:60 kDa heat shock protein, mitochondrial
- Antigen-Symbol:HSPD1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Western blot:Yes
- Epitop:187-215 aa
- Form:Liquid
- Gen-ID:3329
- Modification:None
- Molecular weight:61 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:2 mg/ml
- Transporttemperatur:4 °C
- Immunogen:This HSPD1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 187-215 amino acids from the central region of human HSPD1.
- Reinigung:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- VE:400 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:100.
Type: Primary
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 187-215
Host: Rabbit
Isotype: Ig
Reactivity: Human