Anti-CAMK1G Rabbit Polyclonal Antibody
Catalog # PRSI22-902
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Calcium/calmodulin-dependent protein kinase type 1G
- Antigen-Symbol:CAMK1G
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:57172
- Antigen-Synonyme:VWS1|CAMK1G|CLICK III|CaM-KI gamma|CaMKIG|CaM kinase I gamma|CaM kinase IG|CaMK-like CREB kinase III|CLICK3|CaMKI gamma|Calcium/calmodulin-dependent protein kinase type 1G
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 217-476 of human CAMK1G (NP_065172.1)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: CAMK1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat