Anti-ZDHHC17 Rabbit Polyclonal Antibody
Catalog # PRSI22-490
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Zinc finger, DHHC-type containing 17
- Antigen-Symbol:ZDHHC17
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:23390
- Antigen-Synonyme:KIAA0946|HSPC294|ZDHHC17|HYPH|HIP14|HIP3
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 170-310 of human ZDHHC17 (NP_056151.2)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: ZDHHC17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat